Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human EGFRvIII Protein, His Tag (Site-specific conjugation)
Catalog No. 502107503
Encompass_Preferred
Recombinant Protein;200TESTS ;PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is produced via site-specific conjugation of PE to Human EGFRvIII (25-378), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.5 kDa.
Catalog No. 50-210-7503 Supplier ACROBiosystems Supplier No. EGIHP2E3200TESTS
Recombinant Protein;200TESTS ;PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is produced via site-specific conjugation of PE to Human EGFRvIII (25-378), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.5 kDa.