Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human Glypican 3 / GPC3 Protein, His Tag (Site-specific conjugation)
Catalog No. 502107508
Encompass_Preferred
Recombinant Protein;25TESTS ;PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is produced via site-specific conjugation of PE to Human Glypican 3 (25-559), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 64.5 kDa.
Catalog No. 50-210-7508 Supplier ACROBiosystems Supplier No. GP3HP2E325TESTS
Recombinant Protein;25TESTS ;PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is produced via site-specific conjugation of PE to Human Glypican 3 (25-559), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 64.5 kDa.