Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human GUCY2C / Guanylyl cyclase C Protein, His Tag (Site-specific conjugation)
Supplier:  ACROBiosystems GUCHP2E3200TESTS
Encompass_Preferred
Recombinant Protein;200TESTS ;PE-Labeled Human GUCY2C, His Tag (GUC-HP2E3) is produced via site-specific conjugation of PE to Human EGFRvIII (24-430), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human GUCY2C, His Tag (GUC-HP2E3) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.6 kDa.
Catalog No. 50-210-7501