Learn More
ACROBiosystems PE-Labeled Human HLA-A0201B
Catalog No. 502534758
Encompass_Preferred
PE-Labeled Human HLA-A*0201B2MAFP (FMNKFIYEI) Tetramer Protein (HLA-HP2H5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) FMNKFIYEI peptide (Accession # AAA59606.1 (HLA-A*0201) P61769 (B2M) FMNKFIYEI).
Catalog No. 50-253-4758 Supplier ACROBiosystems Supplier No. HLAHP2H525UG
PE-Labeled Human HLA-A*0201B2MAFP (FMNKFIYEI) Tetramer Protein (HLA-HP2H5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) FMNKFIYEI peptide (Accession # AAA59606.1 (HLA-A*0201) P61769 (B2M) FMNKFIYEI).