Learn More
ACROBiosystems PE-Labeled Human HLA-A*11:01&B2M&KRASG12V (VVVGAVGVGK) Tetramer Protein
Catalog No. 502719378
Encompass_Preferred
The Kirsten rat sarcoma 2 viral oncogene homolog (KRAS) oncogene plays a critical role in the initiation and maintenance of pancreatic tumors and its signaling network represents a major target for therapeutic intervention. The Human HLA-A*1101 KRASG12V (VVVGAVGVGK) complex protein is a complex of HLA-A*1101 of the MHC Class I, B2M, and VVVGAVGVGK peptide of the KRASG12V.
Catalog No. 50-271-9378 Supplier ACROBiosystems Supplier No. HLVHP2H725UG
The Kirsten rat sarcoma 2 viral oncogene homolog (KRAS) oncogene plays a critical role in the initiation and maintenance of pancreatic tumors and its signaling network represents a major target for therapeutic intervention. The Human HLA-A*1101 KRASG12V (VVVGAVGVGK) complex protein is a complex of HLA-A*1101 of the MHC Class I, B2M, and VVVGAVGVGK peptide of the KRASG12V.