Learn More
ACROBiosystems PE-Labeled Human HLA-A*11:01&B2M&KRAS (VVVGAGGVGK) Tetramer Protein
Catalog No. 502719392
Encompass_Preferred
The Kirsten rat sarcoma 2 viral oncogene homolog (KRAS) oncogene plays a critical role in the initiation and maintenance of pancreatic tumors and its signaling network represents a major target for therapeutic intervention. The Alex Fluor 647 labeled Human HLA-A*1101 KRAS (VVVGAGGVGK) complex protein is a complex of HLA-A*1101 of the MHC Class I, B2M, and VVVGAGGVGK peptide of the KRAS.
Catalog No. 50-271-9392 Supplier ACROBiosystems Supplier No. HLSHP2H625UG
The Kirsten rat sarcoma 2 viral oncogene homolog (KRAS) oncogene plays a critical role in the initiation and maintenance of pancreatic tumors and its signaling network represents a major target for therapeutic intervention. The Alex Fluor 647 labeled Human HLA-A*1101 KRAS (VVVGAGGVGK) complex protein is a complex of HLA-A*1101 of the MHC Class I, B2M, and VVVGAGGVGK peptide of the KRAS.