Learn More
ACROBiosystems PE-Labeled Human HLA-A1101B
Catalog No. 502534765
Encompass_Preferred
PE-Labeled Human HLA-A*1101B2MHPV16-E7 (IVCPICSQK) Tetramer Protein (HL7-HP2H4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*1101) Ile 21 - Met 119 (B2M) IVCPICSQK peptide (Accession # Q5S3G3-1 (HLA-A*1101) P61769 (B2M) IVCPICSQK).
Catalog No. 50-253-4765 Supplier ACROBiosystems Supplier No. HL7HP2H4200UG
PE-Labeled Human HLA-A*1101B2MHPV16-E7 (IVCPICSQK) Tetramer Protein (HL7-HP2H4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*1101) Ile 21 - Met 119 (B2M) IVCPICSQK peptide (Accession # Q5S3G3-1 (HLA-A*1101) P61769 (B2M) IVCPICSQK).