Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human Mesothelin / MSLN (296-580) Protein, His Tag (Site-specific conjugation)
Supplier:  ACROBiosystems MSNHP2H525TESTS
Encompass_Preferred
Recombinant Protein;25TESTS ;PE-Labeled Human Mesothelin (296-580), His Tag (MSN-HP2H5) is produced via site-specific conjugation of PE to Human Mesothelin (296-580), His Tag under optimal conditions with a proprietary technology. Human Mesothelin (296-580), His Tag is expressed in HEK293 cells containing AA Glu 296 - Gly 580 (Accession # AAH09272).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.9 kDa. ;1e6 of the anti-MSLN CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human Mesothelin (296-580) Protein, His Tag (Cat. No. MSN-HP2H5) and negative control protein respectively, PE signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-210-7176