Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human PSMA / FOLH1 Protein, His Tag
Supplier:  ACROBiosystems PSAHP2Q3200TESTS
Encompass_Preferred
Recombinant Protein;200TESTS;PE-Labeled Human PSMA, His Tag (PSA-HP2Q3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1 ).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 83.1 kDa.
Catalog No. 50-210-6597