Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human ROR1 Protein, His Tag (Site-specific conjugation)
Catalog No. 502107505
Encompass_Preferred
Recombinant Protein;25TESTS ;PE-Labeled Human ROR1, His Tag (RO1-HP2E3) is produced via site-specific conjugation of PE to Human ROR1 (30-403), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human ROR1, His Tag (RO1-HP2E3) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.6 kDa.
Catalog No. 50-210-7505 Supplier ACROBiosystems Supplier No. RO1HP2E3200TESTS
Recombinant Protein;25TESTS ;PE-Labeled Human ROR1, His Tag (RO1-HP2E3) is produced via site-specific conjugation of PE to Human ROR1 (30-403), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human ROR1, His Tag (RO1-HP2E3) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.6 kDa.