Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human Siglec-3 / CD33 Protein, His Tag (Site-specific conjugation)
Catalog No. 502106366
Encompass_Preferred
Recombinant Protein;200TESTS;PE-Labeled Human Siglec-3, His Tag (CD3-HP2E3) is produced via site-specific conjugation of PE to Human Siglec-3, His Tag under optimal conditions with a proprietary technology. Human Siglec-3, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.4 kDa. ;5e5 of anti-Siglec-3 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human Siglec-3, His Tag (Cat. No. CD3-HP2E3) and negative control protein respectively. PE signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-210-6366 Supplier ACROBiosystems Supplier No. CD3HP2E3200TESTS
Recombinant Protein;200TESTS;PE-Labeled Human Siglec-3, His Tag (CD3-HP2E3) is produced via site-specific conjugation of PE to Human Siglec-3, His Tag under optimal conditions with a proprietary technology. Human Siglec-3, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.4 kDa. ;5e5 of anti-Siglec-3 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human Siglec-3, His Tag (Cat. No. CD3-HP2E3) and negative control protein respectively. PE signal was used to evaluate the binding activity (QC tested).