Learn More
ACROBiosystems PE-Labeled Human SLAMF7 / CRACC / CD319 Protein, His Tag
Catalog No. 502054899
Encompass_Preferred
Recombinant Protein;25TESTS;PE-Labeled Human SLAMF7, His Tag (SL7-HP2H3) is produced via conjugation of PE to Human SLAMF7, His Tag under optimal conditions. Human SLAMF7, His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # Q9NQ25-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. ;1e6 of the anti-SLAMF7 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human SLAMF7, His Tag (Cat. No. SL7-HP2H3) and negative control protein respectively, PE signals was used to evaluate the binding activity (QC tested).
Catalog No. 50-205-4899 Supplier ACROBiosystems Supplier No. SL7HP2H325TESTS
Recombinant Protein;25TESTS;PE-Labeled Human SLAMF7, His Tag (SL7-HP2H3) is produced via conjugation of PE to Human SLAMF7, His Tag under optimal conditions. Human SLAMF7, His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # Q9NQ25-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. ;1e6 of the anti-SLAMF7 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human SLAMF7, His Tag (Cat. No. SL7-HP2H3) and negative control protein respectively, PE signals was used to evaluate the binding activity (QC tested).