Learn More
ACROBiosystems PE-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Y45)
Supplier:  ACROBiosystems FM3HPY5325TESTS
Encompass_Preferred
Antibody;25TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;PE-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-HPY53) is produced via conjugation of PE to Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 under optimal conditions.;This protein . ;1e6 of the Anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Y45) (Cat. No. FM3-HPY53) and negative control antibody respectively, PE signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-205-5008