Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems PE-Labeled Monoclonal Anti-FMC63 Antibody, Rabbit IgG (Clone 1G10), 25 tests

Catalog No. NC3853595
Encompass_Preferred

PE-Labeled monoclonal anti-FMC63 antibody (rabbit IgG, clone 1G10) is a PE-conjugated reagent designed for flow cytometry detection of FMC63-derived CAR (anti-CD19 scFv) expression. Supplied lyophilized with stabilizers and optimized for FACS staining.

  • Label: PE.
  • Host species: Rabbit IgG.
  • Clone: 1G10.
  • Recommended use: Flow cytometry for detection of FMC63-derived CARs.
  • Typical staining: 2 μL per 1x10^6 cells in 100 μL (approx. 1:50).
  • Formulation: PBS, pH 7.4, 0.2% BSA, 0.03% Proclin 300, with trehalose as protectant.
  • Storage: Lyophilized -20°C to -70°C; after reconstitution -70°C or 2-8°C.
  • Available sizes: 25-test and 200-test formats.

Catalog No. NC3853595 Supplier ACROBiosystems Supplier No. FM3PFM72125TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PE-Labeled monoclonal anti-FMC63 antibody (rabbit IgG, clone 1G10) is a PE-conjugated reagent designed for flow cytometry detection of FMC63-derived CAR (anti-CD19 scFv) expression. Supplied lyophilized with stabilizers and optimized for FACS staining.

  • Label: PE.
  • Host species: Rabbit IgG.
  • Clone: 1G10.
  • Recommended use: Flow cytometry for detection of FMC63-derived CARs.
  • Typical staining: 2 μL per 1x10^6 cells in 100 μL (approx. 1:50).
  • Formulation: PBS, pH 7.4, 0.2% BSA, 0.03% Proclin 300, with trehalose as protectant.
  • Storage: Lyophilized -20°C to -70°C; after reconstitution -70°C or 2-8°C.
  • Available sizes: 25-test and 200-test formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.