Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems PE-Labeled Monoclonal Anti-Human CD5 Antibody, Mouse IgG1 (Clone 2C8A7), 50 tests

Catalog No. NC3870782
Encompass_Preferred

This PE-labeled mouse monoclonal antibody (clone 2C8A7) specifically recognizes human CD5 and is formulated for flow cytometry to detect CD5-expressing cells. The reagent is conjugated to phycoerythrin (PE) and supplied for 50 tests per vial with 0.03% Proclin as preservative.

  • Conjugate: PE.
  • Target: human CD5.
  • Host species: mouse; clone 2C8A7.
  • Isotype: Mouse IgG1, kappa.
  • Pack size: 50 tests per vial.
  • Preservative: 0.03% Proclin.
  • Application: Flow cytometry.

Catalog No. NC3870782 Supplier ACROBiosystems Supplier No. FABM0320150TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This PE-labeled mouse monoclonal antibody (clone 2C8A7) specifically recognizes human CD5 and is formulated for flow cytometry to detect CD5-expressing cells. The reagent is conjugated to phycoerythrin (PE) and supplied for 50 tests per vial with 0.03% Proclin as preservative.

  • Conjugate: PE.
  • Target: human CD5.
  • Host species: mouse; clone 2C8A7.
  • Isotype: Mouse IgG1, kappa.
  • Pack size: 50 tests per vial.
  • Preservative: 0.03% Proclin.
  • Application: Flow cytometry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.