Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems PE-Labeled Monoclonal Anti-Human CD73 Antibody, Mouse IgG1 (clone 5A10) - PE conjugate, 100 tests

Catalog No. NC3794382
Encompass_Preferred

PE-conjugated monoclonal antibody targeting human CD73 (clone 5A10), provided as mouse IgG1 and validated for flow cytometry. Recommended use is 1:20 dilution (5 μL per 1x10^6 cells in 100 μL). Supplied as a 100-tests pack and stable for 12 months when stored at 2-8 °C; protect from light.

  • Format: PE-labeled antibody.
  • Clone: 5A10.
  • Isotype/species: Mouse IgG1.
  • Application: Flow cytometry.
  • Recommended dilution: 1:20 (5 μL per 1x10^6 cells in 100 μL).
  • Pack size: 100 tests (50 tests x 2).
  • Excitation/emission: 488 nm / 561 nm excitation; 575 nm emission.
  • Storage: 2-8 °C, stable for 12 months.
  • Shipping: Supplied with blue ice.

Catalog No. NC3794382 Supplier ACROBiosystems Supplier No. FABM02401100TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PE-conjugated monoclonal antibody targeting human CD73 (clone 5A10), provided as mouse IgG1 and validated for flow cytometry. Recommended use is 1:20 dilution (5 μL per 1x10^6 cells in 100 μL). Supplied as a 100-tests pack and stable for 12 months when stored at 2-8 °C; protect from light.

  • Format: PE-labeled antibody.
  • Clone: 5A10.
  • Isotype/species: Mouse IgG1.
  • Application: Flow cytometry.
  • Recommended dilution: 1:20 (5 μL per 1x10^6 cells in 100 μL).
  • Pack size: 100 tests (50 tests x 2).
  • Excitation/emission: 488 nm / 561 nm excitation; 575 nm emission.
  • Storage: 2-8 °C, stable for 12 months.
  • Shipping: Supplied with blue ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.