Learn More
ACROBiosystems PE-Labeled Mouse H-2Kb&B2M&OVA (SIINFEKL) Tetramer Protein
Catalog No. 502719529
Encompass_Preferred
Ovalbumin (OVA) can induce both humoral and cellular immune responses based on well-characterized peptide epitopes. The OVA257-264 octapeptide was one of the first OVA epitopes to be characterized, it has an amino acid sequence SIINFEKL, which is recognized by cytotoxic T lymphocytes. ovalbumin (Ova) was presented to T cells if it was released from pinosomes into the cytosol by osmotic lysis of pinosomes. Immunization with the adjuvanted SIINFEKL peptide induces long-lasting CD8+ T cell immunity in mice.
Catalog No. 50-271-9529 Supplier ACROBiosystems Supplier No. H2AMP2H725UG
Ovalbumin (OVA) can induce both humoral and cellular immune responses based on well-characterized peptide epitopes. The OVA257-264 octapeptide was one of the first OVA epitopes to be characterized, it has an amino acid sequence SIINFEKL, which is recognized by cytotoxic T lymphocytes. ovalbumin (Ova) was presented to T cells if it was released from pinosomes into the cytosol by osmotic lysis of pinosomes. Immunization with the adjuvanted SIINFEKL peptide induces long-lasting CD8+ T cell immunity in mice.