Learn More
ABclonal Technology PE Mouse anti-Human CD42a mAb
SDP
Catalog No. 5000010018
Encompass_Preferred
This gene encodes a small membrane glycoprotein found on the surface of human platelets It forms a 1-to-1 noncovalent complex with glycoprotein Ib a platelet surface membrane glycoprotein complex that functions as a receptor for von Willebrand factor The complete receptor complex includes noncovalent association of the alpha and beta subunits with the protein encoded by this gene and platelet glycoprotein V Defects in this gene are a cause of Bernard-Soulier syndrome also known as giant platelet disease These patients have unusually large platelets and have a clinical bleeding tendency
Catalog No. 50-000-10018 Supplier ABclonal Technology Supplier No. A27731200T
This gene encodes a small membrane glycoprotein found on the surface of human platelets It forms a 1-to-1 noncovalent complex with glycoprotein Ib a platelet surface membrane glycoprotein complex that functions as a receptor for von Willebrand factor The complete receptor complex includes noncovalent association of the alpha and beta subunits with the protein encoded by this gene and platelet glycoprotein V Defects in this gene are a cause of Bernard-Soulier syndrome also known as giant platelet disease These patients have unusually large platelets and have a clinical bleeding tendency