Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PE Mouse Anti-Human IgA1, BD Pharmingen™

Catalog No. BDB571775 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571775 25 Tests
BDB571774 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571775 Supplier BD Biosciences Supplier No. 571775
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The B3506B4 monoclonal antibody specifically recognizes the Fc region of the human Immunoglobulin A (IgA) subclass known as IgA1. IgA1 is comprised of two identical heavy chains encoded by IGHA1, and two light chains, either Igκ or Igλ, that are linked together by disulfide bonds. The B3506B4 antibody does not crossreact with other human immunoglobulin heavy chain (IgH) subclasses, including the IgA2 subclass. IgA1 is expressed by plasmablasts, plasma cells, and memory B cells as well as by some myeloma or plasmacytoma cells. IgA1 is expressed in either a transmembrane form that serves as an antigen-specific B cell surface receptor or in a soluble secreted form. Monomeric and dimeric forms of IgA1 are found in the serum although the monomeric form predominates. Multimeric forms of IgA1 connected by a J-chain and secretory component are also found in bodily secretions. Secretory IgA is transported across epithelial cells and secreted into the lumen of mucosal tissues including the gastrointestinal and respiratory tracts and is found in human breast milk. IgA1 functions in the agglutination of microbes and can block their adherence to or infection of human cells. It can also bind to CD89 (FcαRI) expressed on neutrophils, monocytes, macrophages, and eosinophils to trigger a variety of immune responses including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and the release of inflammatory cytokines and mediators such as in the response to microbes.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone B3506B4 (also known as DLDB4)
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias IGHA1; Immunoglobulin alpha 1
Host Species Mouse
Immunogen Human IgA1 Myeloma Protein
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.