Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PE Mouse Anti-Human IgA2, BD Pharmingen™

Catalog No. BDB571777 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571777 25 Tests
BDB571776 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571777 Supplier BD Biosciences Supplier No. 571777
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The A9604D2 monoclonal antibody specifically recognizes the Fc region of the human Immunoglobulin A (IgA) subclass known as IgA2. It does not cross-react with other human immunoglobulin heavy chain (IgH) subclasses, including the IgA1 subclass. IgA2 is expressed by plasmablasts, plasma cells, and memory B cells, as well as some myeloma or plasmacytoma cells. It can exist as a transmembrane form (antigen-specific B cell surface receptor) or a soluble secreted form. Monomeric and dimeric forms of IgA2 are found in serum, while multimeric forms of secretory IgA2 (connected by a J-chain and secretory component) are typically found in higher levels in bodily secretions and human breast milk. Secretory IgA2 functions in agglutination of microbes and can block their adherence or infection of human cells. It can also bind to CD89 (FcαRI) on neutrophils, monocytes, macrophages, and eosinophils to trigger various immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone A9604D2 (also known as DLDD2)
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias IGHA2; Immunoglobulin alpha 2
Host Species Mouse
Immunogen Human IgA2 Myeloma Protein
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3494
Target Species Human
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.