Learn More
Biolegend PE Mouse IgG2b, κ Isotype Ctrl
Supplier:  Biolegend 402204
Encompass_Preferred
PE Mouse IgG2b, κ Isotype Ctrl [27-35]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC; Size: 100 ug
Catalog No. NC1426881