Learn More
Cytek Biosciences Inc PE aMu CD209b Anti Primary
Supplier: Cytek Biosciences Inc 502093U100
Product Name: PE Anti-Mouse CD209b (SIGN-R1) (22D1)
Product Class: Antibody
Type: Primary
Product Name: PE Anti-Mouse CD209b (SIGN-R1) (22D1)
Cytek Catalog Number: 50-2093-U100
Marker: CD209b (SIGN-R1)
Target Species: Mouse
Clonality: Monoclonal
Clone: 22D1
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: PE
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.