Learn More
Cytek Biosciences Inc PE MS F4/80 (BM8.1) 100UG
Supplier:  Cytek Biosciences Inc 504801U100
Encompass_Preferred
PE Anti-Mouse F4/80 Antigen (BM8.1) Isotype: Rat IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-4759