Learn More
Biolegend PE Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 400112
Encompass_Preferred
PE Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 100 ug
Catalog No. NC9025796