Learn More
Biolegend PE Mouse IgG2a, κ isotype Ctrl
Supplier:  Biolegend 400211
Encompass_Preferred
PE Mouse IgG2a, κ isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Apps: FC, ICFC; Size: 25 ug
Catalog No. NC0176456