Learn More
Biolegend PE Mouse IgG2b, κ isotype Ctrl
Supplier:  Biolegend 400313
Encompass_Preferred
PE Mouse IgG2b, κ isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 25 tests
Catalog No. 50-168-902