Learn More
Biolegend PE Mouse IgG2b, κ isotype Ctrl
Supplier:  Biolegend 400312
Encompass_Preferred
PE Mouse IgG2b, κ isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 100 ug
Catalog No. 50-168-901