Learn More
MSC Vikan Hand Brush / Nailbrush, Hard, 130 mm, Green
Catalog No. 502838439
Encompass
This hand brush/nailbrush is designed for efficient cleaning in various environments, including food processing. It features hard polyester bristles and a robust polypropylene block, making it durable and suitable for demanding tasks. The brush is 130 mm long and colored green, aiding in HACCP compliance.
  • Effective for scrubbing with hard polyester bristles.
  • Durable polypropylene block ensures longevity.
  • Suitable for contact with various food types.
  • Withstands temperatures from -20 °C to 100 °C.
  • Autoclavable up to 121 °C for sterilization.
  • Complies with FDA and EU food contact regulations.
  • Designed for repeated use and easy cleaning.
Catalog No. 50-283-8439 Supplier MSC Supplier No. 52584141
This hand brush/nailbrush is designed for efficient cleaning in various environments, including food processing. It features hard polyester bristles and a robust polypropylene block, making it durable and suitable for demanding tasks. The brush is 130 mm long and colored green, aiding in HACCP compliance.
  • Effective for scrubbing with hard polyester bristles.
  • Durable polypropylene block ensures longevity.
  • Suitable for contact with various food types.
  • Withstands temperatures from -20 °C to 100 °C.
  • Autoclavable up to 121 °C for sterilization.
  • Complies with FDA and EU food contact regulations.
  • Designed for repeated use and easy cleaning.