Learn More
Abcam PE / R-Phycoerythrin Conjugation Kit - Lightning-Link, 5 x 600UG
Catalog No. 502585578
Encompass_Preferred
The Abcam PE/R-Phycoerythrin Conjugation Kit (ab102918) offers several standout features: -Rapid Conjugation: achieve PE/R-Phycoerythrin labeling in under 4 hours with just 30 seconds of hands-on time. -High Efficiency: ensures 100% antibody recovery, preserving the integrity and value of your antibodies. -Versatility: suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence: cited in over 50 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5578 Supplier Abcam Supplier No. AB1029185X600UG
The Abcam PE/R-Phycoerythrin Conjugation Kit (ab102918) offers several standout features: -Rapid Conjugation: achieve PE/R-Phycoerythrin labeling in under 4 hours with just 30 seconds of hands-on time. -High Efficiency: ensures 100% antibody recovery, preserving the integrity and value of your antibodies. -Versatility: suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence: cited in over 50 publications
The product is subject to the following: Abcam Restricted Use Statement