Learn More
ABclonal Technology PE Rabbit anti- CD150/SLAM
SDP
Catalog No. 5000010175
Encompass_Preferred
Enables identical protein binding activity and signaling receptor activity Involved in natural killer cell activation positive regulation of activated T cell proliferation and regulation of cytokine production Acts upstream of or within several processes including leukocyte chemotaxis involved in inflammatory response positive regulation of leukocyte chemotaxis and regulation of vesicle fusion Located in external side of plasma membrane and phagocytic vesicle Is expressed in liver lobe Orthologous to human SLAMF1 (signaling lymphocytic activation molecule family member 1)
Catalog No. 50-000-10175 Supplier ABclonal Technology Supplier No. A26519500T
Enables identical protein binding activity and signaling receptor activity Involved in natural killer cell activation positive regulation of activated T cell proliferation and regulation of cytokine production Acts upstream of or within several processes including leukocyte chemotaxis involved in inflammatory response positive regulation of leukocyte chemotaxis and regulation of vesicle fusion Located in external side of plasma membrane and phagocytic vesicle Is expressed in liver lobe Orthologous to human SLAMF1 (signaling lymphocytic activation molecule family member 1)