Learn More
ABclonal Technology PE Rabbit anti- CD262/TRAILR2
SDP
Catalog No. 5000011250
Encompass_Preferred
Predicted to enable TRAIL receptor activity identical protein binding activity and protease binding activity Predicted to be involved in TRAIL-activated apoptotic signaling pathway and positive regulation of apoptotic process Predicted to act upstream of or within apoptotic process and regulation of apoptotic process Predicted to be located in Golgi apparatus cytosol and membrane raft Predicted to be active in cell surface and plasma membrane Is expressed in bladder liver renal vasculature urethra of female and urethra of male Human ortholog(s) of this gene implicated in carcinoma (multiple) cervical cancer hematologic cancer (multiple) and urinary bladder cancer Orthologous to several human genes including TNFRSF10A (TNF receptor superfamily member 10a)
Catalog No. 50-000-11250 Supplier ABclonal Technology Supplier No. A24798100T
Predicted to enable TRAIL receptor activity identical protein binding activity and protease binding activity Predicted to be involved in TRAIL-activated apoptotic signaling pathway and positive regulation of apoptotic process Predicted to act upstream of or within apoptotic process and regulation of apoptotic process Predicted to be located in Golgi apparatus cytosol and membrane raft Predicted to be active in cell surface and plasma membrane Is expressed in bladder liver renal vasculature urethra of female and urethra of male Human ortholog(s) of this gene implicated in carcinoma (multiple) cervical cancer hematologic cancer (multiple) and urinary bladder cancer Orthologous to several human genes including TNFRSF10A (TNF receptor superfamily member 10a)