Learn More
ABclonal Technology PE Rabbit anti-Human CD11a mAb
SDP
Catalog No. 5000013239
Encompass_Preferred
ITGAL encodes the integrin alpha L chain Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain This I-domain containing alpha integrin combines with the beta 2 chain (ITGB2) to form the integrin lymphocyte function-associated antigen-1 (LFA-1) which is expressed on all leukocytes LFA-1 plays a central role in leukocyte intercellular adhesion through interactions with its ligands ICAMs 1-3 (intercellular adhesion molecules 1 through 3) and also functions in lymphocyte costimulatory signaling Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-13239 Supplier ABclonal Technology Supplier No. A26540100T
ITGAL encodes the integrin alpha L chain Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain This I-domain containing alpha integrin combines with the beta 2 chain (ITGB2) to form the integrin lymphocyte function-associated antigen-1 (LFA-1) which is expressed on all leukocytes LFA-1 plays a central role in leukocyte intercellular adhesion through interactions with its ligands ICAMs 1-3 (intercellular adhesion molecules 1 through 3) and also functions in lymphocyte costimulatory signaling Two transcript variants encoding different isoforms have been found for this gene