Learn More
ABclonal Technology PE Rabbit anti-Human CD138 mAb
SDP
Catalog No. 5000013847
Encompass_Preferred
The protein encoded by this gene is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family The syndecans mediate cell binding cell signaling and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein The syndecan-1 protein functions as an integral membrane protein and participates in cell proliferation cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins Altered syndecan-1 expression has been detected in several different tumor types While several transcript variants may exist for this gene the full-length natures of only two have been described to date These two represent the major variants of this gene and encode the same protein
Catalog No. 50-000-13847 Supplier ABclonal Technology Supplier No. A27777500T
The protein encoded by this gene is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family The syndecans mediate cell binding cell signaling and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein The syndecan-1 protein functions as an integral membrane protein and participates in cell proliferation cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins Altered syndecan-1 expression has been detected in several different tumor types While several transcript variants may exist for this gene the full-length natures of only two have been described to date These two represent the major variants of this gene and encode the same protein