Learn More
ABclonal Technology PE Rabbit anti-Human CD22 mAb
SDP
Catalog No. 5000014134
Encompass_Preferred
Predicted to enable CD4 receptor binding activity protein phosphatase binding activity and sialic acid binding activity Involved in B cell activation negative regulation of B cell receptor signaling pathway and regulation of endocytosis Located in early endosome and recycling endosome
Catalog No. 50-000-14134 Supplier ABclonal Technology Supplier No. A27750500T
Predicted to enable CD4 receptor binding activity protein phosphatase binding activity and sialic acid binding activity Involved in B cell activation negative regulation of B cell receptor signaling pathway and regulation of endocytosis Located in early endosome and recycling endosome