Learn More
ABclonal Technology PE Rabbit anti-Human CD23 mAb
SDP
Catalog No. 5000009180
Encompass_Preferred
The protein encoded by this gene is a B-cell specific antigen and a low-affinity receptor for IgE It has essential roles in B cell growth and differentiation and the regulation of IgE production This protein also exists as a soluble secreted form then functioning as a potent mitogenic growth factor Alternatively spliced transcript variants encoding different isoforms have been described for this gene
Catalog No. 50-000-09180 Supplier ABclonal Technology Supplier No. A26174500T
The protein encoded by this gene is a B-cell specific antigen and a low-affinity receptor for IgE It has essential roles in B cell growth and differentiation and the regulation of IgE production This protein also exists as a soluble secreted form then functioning as a potent mitogenic growth factor Alternatively spliced transcript variants encoding different isoforms have been described for this gene