Learn More
ABclonal Technology PE Rabbit anti-Human CD44 mAb
Supplier: ABclonal Technology A24419100T
The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions cell adhesion and migration It is a receptor for hyaluronic acid (HA) and can also interact with other ligands such as osteopontin collagens and matrix metalloproteinases (MMPs) This protein participates in a wide variety of cellular functions including lymphocyte activation recirculation and homing hematopoiesis and tumor metastasis Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms however the full length nature of some of these variants has not been determined Alternative splicing is the basis for the structural and functional diversity of this protein and may be related to tumor metastasis