Learn More
ABclonal Technology PE Rabbit anti-Human CD6 mAb
SDP
Catalog No. 5000012681
Encompass_Preferred
This gene encodes a protein found on the outer membrane of T-lymphocytes as well as some other immune cells The encoded protein contains three scavenger receptor cysteine-rich (SRCR) domains and a binding site for an activated leukocyte cell adhesion molecule The gene product is important for continuation of T cell activation This gene may be associated with susceptibility to multiple sclerosis (PMID 19525953 21849685) Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12681 Supplier ABclonal Technology Supplier No. A26536200T
This gene encodes a protein found on the outer membrane of T-lymphocytes as well as some other immune cells The encoded protein contains three scavenger receptor cysteine-rich (SRCR) domains and a binding site for an activated leukocyte cell adhesion molecule The gene product is important for continuation of T cell activation This gene may be associated with susceptibility to multiple sclerosis (PMID 19525953 21849685) Multiple transcript variants encoding different isoforms have been found for this gene