Learn More
ABclonal Technology PE Rabbit anti-Human CD7 mAb
Supplier: ABclonal Technology A27376500T
This gene encodes a transmembrane protein which is a member of the immunoglobulin superfamily This protein is found on thymocytes and mature T cells It plays an essential role in T-cell interactions and also in T-cell B-cell interaction during early lymphoid development