Learn More
ABclonal Technology PE Rabbit anti-Human CD80/B7-1
SDP
Catalog No. 5000013621
Encompass_Preferred
The protein encoded by this gene is a membrane receptor that is activated by the binding of CD28 or CTLA-4 The activated protein induces T-cell proliferation and cytokine production This protein can act as a receptor for adenovirus subgroup B and may play a role in lupus neuropathy
Catalog No. 50-000-13621 Supplier ABclonal Technology Supplier No. A27778200T
The protein encoded by this gene is a membrane receptor that is activated by the binding of CD28 or CTLA-4 The activated protein induces T-cell proliferation and cytokine production This protein can act as a receptor for adenovirus subgroup B and may play a role in lupus neuropathy