Learn More
ABclonal Technology PE Rabbit anti-Human CPT1A mAb
SDP
Catalog No. 5000012486
Encompass_Preferred
The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable) together with a carnitine-acylcarnitine translocase CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12486 Supplier ABclonal Technology Supplier No. A27106100T
The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable) together with a carnitine-acylcarnitine translocase CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation Alternatively spliced transcript variants encoding different isoforms have been found for this gene