Learn More
ABclonal Technology PE Rabbit anti-Human CR2/CD21
Supplier: ABclonal Technology A25702500T
This gene encodes a membrane protein which functions as a receptor for Epstein-Barr virus (EBV) binding on B and T lymphocytes Genetic variations in this gene are associated with susceptibility to systemic lupus erythematosus type 9 (SLEB9) Alternatively spliced transcript variants encoding different isoforms have been found for this gene