Learn More
ABclonal Technology PE Rabbit anti-Human EGFR mAb
SDP
Supplier:  ABclonal Technology A25699500T
Encompass_Preferred
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily This protein is a receptor for members of the epidermal growth factor family EGFR is a cell surface protein that binds to epidermal growth factor Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation Mutations in this gene are associated with lung cancer
Catalog No. 50-000-12579