Learn More
ABclonal Technology PE Rabbit anti-Human/ FOXP3
Supplier: ABclonal Technology A27686200T
The protein encoded by this gene is a member of the forkhead winged-helix family of transcriptional regulators Defects in this gene are the cause of immunodeficiency polyendocrinopathy enteropathy X-linked syndrome (IPEX) also known as X-linked autoimmunity-immunodeficiency syndrome Alternatively spliced transcript variants encoding different isoforms have been identified