Learn More
ABclonal Technology PE Rabbit anti-Human IFN-gamma
SDP
Catalog No. 5000012499
Encompass_Preferred
This gene encodes a soluble cytokine that is a member of the type II interferon class The encoded protein is secreted by cells of both the innate and adaptive immune systems The active protein is a homodimer that binds to the interferon gamma receptor which triggers a cellular response to viral and microbial infections Mutations in this gene are associated with an increased susceptibility to viral bacterial and parasitic infections and to several autoimmune diseases
Catalog No. 50-000-12499 Supplier ABclonal Technology Supplier No. A27139100T
This gene encodes a soluble cytokine that is a member of the type II interferon class The encoded protein is secreted by cells of both the innate and adaptive immune systems The active protein is a homodimer that binds to the interferon gamma receptor which triggers a cellular response to viral and microbial infections Mutations in this gene are associated with an increased susceptibility to viral bacterial and parasitic infections and to several autoimmune diseases