Learn More
ABclonal Technology PE Rabbit anti-Human IgA1 mAb
SDP
Supplier:  ABclonal Technology A26802100T
Encompass_Preferred
Ig alpha is the major immunoglobulin class in body secretions It may serve both to defend against local infection and to prevent access of foreign antigens to the general immunologic system In normal adult serum the approximate percentage composition of IgA with respect to is subclasses is IgA1 90% and IgA2 10% In secretory IgA the subclass proportions may approach 50 50 The clinical significance of the subclasses has yet to be determined
Catalog No. 50-000-08683