Learn More
ABclonal Technology PE Rabbit anti-Human Ki67 mAb
SDP
Supplier:  ABclonal Technology A27749100T
Encompass_Preferred
Enables protein C-terminus binding activity Involved in regulation of chromosome segregation and regulation of mitotic nuclear division Located in chromosome nuclear body and nucleolus Colocalizes with condensed chromosome Implicated in Crohn s disease breast cancer human immunodeficiency virus infectious disease and pancreatic cancer Biomarker of several diseases including Barrett s esophagus autoimmune disease of musculoskeletal system (multiple) endocrine gland cancer (multiple) gastrointestinal system cancer (multiple) and interstitial cystitis
Catalog No. 50-000-13850