Learn More
ABclonal Technology PE Rabbit anti-Human MICA/MICB
SDP
Catalog No. 5000009650
Encompass_Preferred
This gene encodes a heavily glycosylated protein which is a ligand for the NKG2D type II receptor Binding of the ligand activates the cytolytic response of natural killer (NK) cells CD8 alphabeta T cells and gammadelta T cells which express the receptor This protein is stress-induced and is similar to MHC class I molecules however it does not associate with beta-2-microglobulin or bind peptides Alternative splicing results in multiple transcript variants
Catalog No. 50-000-09650 Supplier ABclonal Technology Supplier No. A27428100T
Add to Cart
Add to Cart
This gene encodes a heavily glycosylated protein which is a ligand for the NKG2D type II receptor Binding of the ligand activates the cytolytic response of natural killer (NK) cells CD8 alphabeta T cells and gammadelta T cells which express the receptor This protein is stress-induced and is similar to MHC class I molecules however it does not associate with beta-2-microglobulin or bind peptides Alternative splicing results in multiple transcript variants