Learn More
ABclonal Technology PE Rabbit anti-Human MICA/MICB
Supplier: ABclonal Technology A27428200T
This gene encodes a heavily glycosylated protein which is a ligand for the NKG2D type II receptor Binding of the ligand activates the cytolytic response of natural killer (NK) cells CD8 alphabeta T cells and gammadelta T cells which express the receptor This protein is stress-induced and is similar to MHC class I molecules however it does not associate with beta-2-microglobulin or bind peptides Alternative splicing results in multiple transcript variants