Learn More
ABclonal Technology PE Rabbit anti-Human NT5E/CD73
SDP
Supplier:  ABclonal Technology A26651200T
Encompass_Preferred
The protein encoded by this gene is a plasma membrane protein that catalyzes the conversion of extracellular nucleotides to membrane-permeable nucleosides The encoded protein is used as a determinant of lymphocyte differentiation Defects in this gene can lead to the calcification of joints and arteries Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12889