Learn More
ABclonal Technology PE Rabbit anti-Human TIGIT mAb
SDP
Catalog No. 5000008824
Encompass_Preferred
This gene encodes a member of the PVR (poliovirus receptor) family of immunoglobin proteins The product of this gene is expressed on several classes of T cells including follicular B helper T cells (TFH) The protein has been shown to bind PVR with high affinity this binding is thought to assist interactions between TFH and dendritic cells to regulate T cell dependent B cell responses
Catalog No. 50-000-08824 Supplier ABclonal Technology Supplier No. A26744200T
This gene encodes a member of the PVR (poliovirus receptor) family of immunoglobin proteins The product of this gene is expressed on several classes of T cells including follicular B helper T cells (TFH) The protein has been shown to bind PVR with high affinity this binding is thought to assist interactions between TFH and dendritic cells to regulate T cell dependent B cell responses